1,4-Dibromobenzene, 99%
1,4-Dibromobenzene, 99%
1,4-Dibromobenzene, 99%
1,4-Dibromobenzene, 99%
Thermo Scientific Chemicals
1,4-Dibromobenzene, 99%
CAS: 106-37-6 | C6H4Br2 | 235.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA112732500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 4-Dibromobenzene
CAS106-37-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0