1,3-Diaminopropane, 99%
1,3-Diaminopropane, 99%
1,3-Diaminopropane, 99%
1,3-Diaminopropane, 99%
Thermo Scientific Chemicals
1,3-Diaminopropane, 99%
CAS: 109-76-2 | C3H10N2 | 74.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
1 L
Packaging:
Glass bottle
Catalog number FSA112352500
Price (MYR)
370.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
370.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 3-Diaminopropane
CAS109-76-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0