1,4-Diaminobutane, 99%
1,4-Diaminobutane, 99%
1,4-Diaminobutane, 99%
1,4-Diaminobutane, 99%
Thermo Scientific Chemicals
1,4-Diaminobutane, 99%
CAS: 110-60-1 | C4H12N2 | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA112121000
Price (MYR)
396.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
396.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 4-Diaminobutane
Name Note99%
CAS110-60-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger