3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%, contains up to 10% water
CAS: 868272-85-9 | C12H18Cl4N4 | 360.10 g/mol
Have Questions?
Change view
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA112090250
Price (MYR)
1,989.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,989.00
BOT
Add to cart
Chemical Identifiers
CAS868272-85-9, 868272-85-9
Specifications
Chemical Name or Material3, 3'-Diaminobenzidine tetrahydrochloride hydrate
CAS868272-85-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0