Decane, 99+%, pure
Decane, 99+%, pure
Decane, 99+%, pure
Thermo Scientific Chemicals
Decane, 99+%, pure
Decane, >-99.0%, C10H22, CAS Number-124-18-5, ccris 653, decane, analytical standard, decyl hydride, dsstox_cid_4913, dsstox_gsid_24913, dsstox_rid_77577, hsdb 63, n-decane, nonane, methyl, unii-nk85062oiy, 2.5L, 99+%, 01, 168, CHEBI:41808, Colorless, 0.7350g/mL, 204-686-4, 142.28, Pure, Danger | CAS: 124-18-5 | C10H22 | 142.286 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number FSA111875000
Price (MYR)
759.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
759.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDecane
Name Note99+%
CAS124-18-5
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.