Cyclohexanemethanol, 99%
Cyclohexanemethanol, 99%
Cyclohexanemethanol, 99%
Thermo Scientific Chemicals
Cyclohexanemethanol, 99%
CAS: 100-49-2 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA111340250
Price (MYR)
462.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
462.00
Each
Specifications
Chemical Name or MaterialCyclohexanemethanol
CAS100-49-2
Density0.9100g/mL
EINECS Number202-857-8
Boiling Point181.0°C