4-Cyanopyridine, 98%
4-Cyanopyridine, 98%
4-Cyanopyridine, 98%
4-Cyanopyridine, 98%
Thermo Scientific Chemicals
4-Cyanopyridine, 98%
CAS: 100-48-1 | C6H4N2 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
View all
Plastic bottle
Catalog number FSA110871000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Cyanopyridine
Name Note98%
CAS100-48-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning