Cinnamyl bromide, 97%, predominantly trans
Cinnamyl bromide, 97%, predominantly trans
Cinnamyl bromide, 97%, predominantly trans
Cinnamyl bromide, 97%, predominantly trans
Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans
CAS: 4392-24-9 | C9H9Br | 197.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA110401000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCinnamyl bromide
Name Note97%
CAS4392-24-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: