Cinnamoyl chloride, 98%, predominantly trans
Cinnamoyl chloride, 98%, predominantly trans
Cinnamoyl chloride, 98%, predominantly trans
Cinnamoyl chloride, 98%, predominantly trans
Thermo Scientific Chemicals
Cinnamoyl chloride, 98%, predominantly trans
CAS: 102-92-1 | C9H7ClO | 166.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA110390050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCinnamoyl chloride
Name Note98%
CAS102-92-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.