3-Chlorophenol, 99%
3-Chlorophenol, 99%
3-Chlorophenol, 99%
Thermo Scientific Chemicals
3-Chlorophenol, 99%
CAS: 108-43-0 | C6H5ClO | 128.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA109811000
Price (MYR)
579.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
579.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Chlorophenol
Name Note99%
CAS108-43-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning