4-Chloro-3-methylphenol, 99+%
4-Chloro-3-methylphenol, 99+%
4-Chloro-3-methylphenol, 99+%
4-Chloro-3-methylphenol, 99+%
Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%
CAS: 59-50-7 | C7H7ClO | 142.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Catalog number FSA109485000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Chloro-3-methylphenol
Name Note99+%
CAS59-50-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger