2-Chlorobutane, 99+%
2-Chlorobutane, 99+%
2-Chlorobutane, 99+%
2-Chlorobutane, 99+%
Thermo Scientific Chemicals
2-Chlorobutane, 99+%
CAS: 78-86-4 | C4H9Cl | 92.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA108832500
Price (MYR)
341.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
341.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Chlorobutane
Name Note99+%
CAS78-86-4
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger