Carbamylcholine chloride, 98+%
Carbamylcholine chloride, 98+%
Carbamylcholine chloride, 98+%
Carbamylcholine chloride, 98+%
Thermo Scientific Chemicals
Carbamylcholine chloride, 98+%
CAS: 51-83-2 | C6H15ClN2O2 | 182.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA108240050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCarbamylcholine chloride
Melting Point200°C to 204°C
CAS51-83-2
ColorWhite
Health Hazard 1GHS Signal Word: Danger