1-Butanol, 99%, extra pure
1-Butanol, 99%, extra pure
1-Butanol, 99%, extra pure
1-Butanol, 99%, extra pure
Thermo Scientific Chemicals
1-Butanol, 99%, extra pure
1-Butanol, 99%, C4H10O, CAS Number-71-36-3, 1-butanol, 1-hydroxybutane, butanol, butyl alcohol, butyl hydroxide, methylolpropane, n-butanol, n-butyl alcohol, propylcarbinol, propylmethanol, 25L, Vapor Pressure: 6.7mbar at 20 deg.C, 01, 367, 117.6 deg.C, 100.0, 98.5, CHEBI:28885, Undesignated | CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA107690025
Price (MYR)
661.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
661.00
EA
Add to cart
Specifications
Chemical Name or Material1-Butanol
Melting Point-89.0°C
CAS71-36-3
CAS Min %98.5
Health Hazard 1Danger