1-Bromonaphthalene, 96%
1-Bromonaphthalene, 96%
1-Bromonaphthalene, 96%
1-Bromonaphthalene, 96%
Thermo Scientific Chemicals
1-Bromonaphthalene, 96%
CAS: 90-11-9 | C10H7Br | 207.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA107155000
Price (MYR)
2,024.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,024.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Bromonaphthalene
Name Note96%
CAS90-11-9
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning