1-Bromohexane, 99+%
1-Bromohexane, 99+%
1-Bromohexane, 99+%
Thermo Scientific Chemicals
1-Bromohexane, 99+%
CAS: 111-25-1 | C6H13Br | 165.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA107045000
Price (MYR)
448.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
448.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Bromohexane
Acidity0.1% max. (as HBr)
CAS111-25-1
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning