1-Bromododecane, 98%
1-Bromododecane, 98%
1-Bromododecane, 98%
1-Bromododecane, 98%
Thermo Scientific Chemicals
1-Bromododecane, 98%
CAS: 143-15-7 | C12H25Br | 249.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA106912500
Price (MYR)
298.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
298.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Bromododecane
CAS143-15-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0