2-Bromobutane, 99+%
2-Bromobutane, 99+%
2-Bromobutane, 99+%
Thermo Scientific Chemicals
2-Bromobutane, 99+%
CAS: 78-76-2 | C4H9Br | 137.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA106781000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromobutane
Name Note99+%
CAS78-76-2
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger