Benzoylhydrazine, 98%
Benzoylhydrazine, 98%
Benzoylhydrazine, 98%
Benzoylhydrazine, 98%
Thermo Scientific Chemicals
Benzoylhydrazine, 98%
CAS: 613-94-5 | C7H8N2O | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA105775000
Price (MYR)
1,833.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,833.00
EA
Add to cart
Specifications
Chemical Name or MaterialBenzoylhydrazine
Name Note98%
CAS613-94-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning