alpha-Benzoin oxime, 98%
alpha-Benzoin oxime, 98%
alpha-Benzoin oxime, 98%
alpha-Benzoin oxime, 98%
Thermo Scientific Chemicals
alpha-Benzoin oxime, 98%
CAS: 441-38-3 | C14H13NO2 | 227.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA105530250
Price (MYR)
306.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
306.00
BOT
Add to cart
Specifications
Chemical Name or Materialα-Benzoin oxime
CAS441-38-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0