Benzethonium chloride, 97%
Benzethonium chloride, 97%
Benzethonium chloride, 97%
Benzethonium chloride, 97%
Thermo Scientific Chemicals
Benzethonium chloride, 97%
Benzethonium chloride, 97%, C27H42ClNO2, CAS Number-121-54-0, benzethoniumchloride, benzethonium chloride, disilyn, hyamine, kylacol, phemeride, phemerol chloride, phemithyn, quatrachlor, solamin, 2.5kg, 96% min. (on dry substance) (Argentometry), 100.0, 96.0, CHEBI:31264, White, 204-479-9, 01,433 | CAS: 121-54-0 | C27H42ClNO2 | 448.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA105385000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzethonium chloride
Name Note97%
CAS121-54-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger