4-Aminosalicylic acid, 99%
4-Aminosalicylic acid, 99%
4-Aminosalicylic acid, 99%
4-Aminosalicylic acid, 99%
Thermo Scientific Chemicals
4-Aminosalicylic acid, 99%
CAS: 65-49-6 | C7H7NO3 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Catalog number FSA104621000
Price (MYR)
404.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
404.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Aminosalicylic acid
Name Note99%
CAS65-49-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning