2-Aminopyridine, 99+%
2-Aminopyridine, 99+%
2-Aminopyridine, 99+%
2-Aminopyridine, 99+%
Thermo Scientific Chemicals
2-Aminopyridine, 99+%
CAS: 504-29-0 | C5H6N2 | 94.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA104541000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Aminopyridine
Name Notep.a.
CAS504-29-0
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger