2-(2-Aminoethyl)pyridine, 95%
2-(2-Aminoethyl)pyridine, 95%
2-(2-Aminoethyl)pyridine, 95%
2-(2-Aminoethyl)pyridine, 95%
Thermo Scientific Chemicals
2-(2-Aminoethyl)pyridine, 95%
CAS: 2706-56-1 | C7H10N2 | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number FSA103690100
Price (MYR)
559.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
559.00
BOT
Add to cart
Specifications
Chemical Name or Material2-(2-Aminoethyl)pyridine
CAS2706-56-1
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0