2-Amino-5-chlorobenzoxazole, 97%
2-Amino-5-chlorobenzoxazole, 97%
2-Amino-5-chlorobenzoxazole, 97%
2-Amino-5-chlorobenzoxazole, 97%
Thermo Scientific Chemicals
2-Amino-5-chlorobenzoxazole, 97%
CAS: 61-80-3 | C7H5ClN2O | 168.58 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA1033800505 gGlass bottle
Catalog number FSA103380050
Price (MYR)
477.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
477.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Amino-5-chlorobenzoxazole
Name Note97%
CAS61-80-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger