Allyl chloride, 98%, stabilized
Allyl chloride, 98%, stabilized
Allyl chloride, 98%, stabilized
Allyl chloride, 98%, stabilized
Thermo Scientific Chemicals
Allyl chloride, 98%, stabilized
CAS: 107-05-1 | C3H5Cl | 76.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA102911000
Price (MYR)
174.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
174.00
EA
Add to cart
Specifications
Chemical Name or MaterialAllyl chloride
CAS107-05-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Alkylation reagent
RUO – Research Use Only