L-Alanine, 99%
L-Alanine, 99%
L-Alanine, 99%
Thermo Scientific Chemicals
L-Alanine, 99%
CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic Bottle
Catalog number FSA102835000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Alanine
Melting Point314.0°C to 316.0°C
Ammonium200ppm max.
CAS56-41-7
pH5.5 to 7 (2.5% soln. at 25°C)