Acetophenone, 98%, pure
Acetophenone, 98%, pure
Acetophenone, 98%, pure
Acetophenone, 98%, pure
Thermo Scientific Chemicals
Acetophenone, 98%, pure
CAS: 98-86-2 | C8H8O | 120.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA102412500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetophenone
CAS98-86-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0