Model 50 Sonic Dismembrator
Model 50 Sonic Dismembrator
Actual product may vary
Model 50 Sonic Dismembrator
Economical, small volume cell disruptor.
Have Questions?
Catalog NumberIncludesElectrical Requirements
FQSFB50220Generator, converter,1/8 in. dia. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included220/230 V, 50/60 Hz
Catalog number FQSFB50220
Price (MYR)
-
Includes:
Generator, converter,1/8 in. dia. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
Electrical Requirements:
220/230 V, 50/60 Hz
The Fisher Scientific™ Model 50 Sonic Dismembrator is compact, portable and extremely simple to operate. Weighing less than 4 lb., this model is the smallest unit on the market and is highly effective for cell disruption, sample preparation and many other small volume applications.
  • Processes 0.2 to 50mL
  • 50 watts
  • Continuous or Remote operation (footswitch optional)
  • Available accessories include: 3 Probe sizes for individual samples
  • Lightweight converter for handheld operation
  • Basic small volume applications
  • Models FB50110 and FB50220 include 1/8 in. probe for processing 0.5-15ml volumes
Specifications
Voltage220/230 V
Electrical Requirements220/230 V, 50/60 Hz
Hertz20 kHz (Ultrasonic Frequency)
Wattage50 W
Volume Range (Metric)0.2 to 50 mL
Dimensions (L x W x H)7 x 8 x 5 in. (17.78 x 20.32 x 12.7 cm)
Height (English)5 in.
Height (Metric)12.7 cm
Length (English)7 in.
Length (Metric)17.78 cm
Width (English)8 in.
Width (Metric)20.32 cm
For Use With (Application)Ideal for basic cell disruption
IncludesGenerator, converter,1/8 in. dia. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
ModelFB50
Unit SizeEach