Replacement Tips for Standard Sonic Dismembrator Probes, Models Q500 and Q700
Replacement Tips for Standard Sonic Dismembrator Probes, Models Q500 and Q700
Actual product may vary
Replacement Tips for Standard Sonic Dismembrator Probes, Models Q500 and Q700
Replace eroded tips easily
Have Questions?
Catalog NumberDiameter (English)Diameter (Metric)
FQSFB44060.5 in.1.27 cm
Catalog number FQSFB4406
Price (MYR)
-
Diameter (English):
0.5 in.
Diameter (Metric):
1.27 cm
  • Replacement Tips for Model 505/705 Sonic Dismembrator
Specifications
Product TypeReplacement Tip
Volume Range (Metric)20 to 250 mL
Diameter (English)0.5 in.
Diameter (English) Tip0.5 in.
Diameter (Metric)1.27 cm
Diameter (Metric) Tip13 mm
For Use With (Equipment)0.5 in. probe for Models Q500 and Q700 sonic dismembrators (FB4220)
Packaging Quantity1 Ea.
Unit SizeEach