Owl™ Blotting Paper
Owl™ Blotting Paper
Actual product may vary
Owl™ Blotting Paper
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with this high-quality blotting paper.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Width (Metric)
FP-410 cm10 cm
FP-120 cm20 cm
FP-235 cm45 cm
Catalog number FP-4
Price (MYR)
489.00
EA
Add to cart
Length (Metric):
10 cm
Width (Metric):
10 cm
Price (MYR)
489.00
EA
Add to cart
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with Thermo Scientific™ Owl™ Blotting Paper, high-quality blotting paper ideal for wicking and blotting applications.
  • High purity, 100% cotton fiber
  • Available in pre-cut mini gel and large format gel sizes
  • Compatible with Owl electroblotting systems
Ordering Information:
  • Sold in packs of 100 sheets
Specifications
Length (Metric)10 cm
Width (Metric)10 cm
Packaging TypeSheets
Flow Rate35 mL/min.
For Use With (Equipment)Owl electroblotting systems
Weight183 g/m2
FormatSheet
Material100% Cotton Fiber
Dimensions (LxW)10 cm x 10 cm
Thickness0.83 mm
Product LineOwl
Unit SizeEA