Barnstead™ Deionization System Disposable Filters
Barnstead™ Deionization System Disposable Filters
Actual product may vary
Barnstead™ Deionization System Disposable Filters
Barnstead™ Deionization System Disposable Filters
Barnstead™ Deionization System Disposable Filters
Extend the life of your Thermo Scientific™ Barnstead™ deionization water system with replacement cartridges and filters.
Have Questions?
Catalog NumberFor Use With (Equipment)Flow RateDescription
FL703X2Easypure;Reverse osmosis and TII water purification system1Final Filter, 0.2 micron, autoclavable; Includes filling bell
Catalog number FL703X2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
Easypure;Reverse osmosis and TII water purification system
Flow Rate:
1
Description:
Final Filter, 0.2 micron, autoclavable; Includes filling bell
Accessory/replacement consumable for Nanopure™ (Analytical, Analytical UV, Biological UF, Life Science UV/UF), Easypure™ II (UV, UF, UV/UF), Easypure II RF (UV), Easypure RoDi, E-Pure™ (3,4 holder), TII.,
Accessory/replacement consumable for Nanopure™ (Analytical, Analytical UV, Biological UF, Life Science UV/UF), Easypure™ II (UV, UF, UV/UF), Easypure II RF (UV), Easypure RoDi, E-Pure™ (3,4 holder), TII.
Specifications
AutoclavableAutoclavable
DescriptionFinal Filter, 0.2 micron, autoclavable; Includes filling bell
Flow Rate1
Pore Size0.2 μm
For Use With (Equipment)Easypure;Reverse osmosis and TII water purification system
IncludesFilling bell
TypeFilter
Unit SizeEA