Check Order Status
Dani 22 Waste Container
Actual product may vary
Fisherbrand
Dani 22 Waste Container
Sharpsguard™ Dani Containers for the safe collection and transportation of broken glass and crockery.
Have Questions?
Catalog Number
Capacity (Metric)
FKS17744874
21.01 L
Catalog number
FKS17744874
Price (MYR)
-
Capacity (Metric):
21.01 L
Sharpsguard™ eco dani 22 is for the safe collection and transportation of broken glass and crockery.
Color-coded with a solid orange lid, handle and label, making it easy to distinguish from other waste containers.
It should be disposed of in landfill sites and therefore is not suitable for hazardous or infectious items.
Specifications
Capacity (Metric)
21.01 L
Diameter (Metric) Outer
324 mm
Color
Orange
Height (Metric)
343 mm
Quantity
1 Ea.
For Use With (Application)
Disposal of Broken Glass and Crockery