PCR™ Cooler, 96-Well, 2 Pack
PCR™ Cooler, 96-Well, 2 Pack
Actual product may vary
PCR™ Cooler, 96-Well, 2 Pack
PCR™ Cooler, 96-Well, 2 Pack
PCR™ Cooler, 96-Well, 2 Pack
PCR™ Cooler, 96-Well, 2 Pack
PCR™ Cooler, 96-Well, 2 Pack
PCR™ Cooler, 96-Well, 2 Pack
PCR™ Cooler, 96-Well, 2 Pack
PCR™ Cooler, 96-Well, 2 Pack
The PCR™ Cooler is ideal for thermosensitive applications, PCR, or cell-based assays, as well as gentle sample thawing and stopping of reactions. The unit also provides a dependable cooling station for pipetting and sample storage. Perfect for transporting samples and reactions between preparation, thermal cycling and post-PCR procedure areas.
Have Questions?
Catalog NumberProduct Type
FKS16320412Purple and Pink and Dark Blue and Light Blue
Catalog number FKS16320412
Price (MYR)
1,280.00
PK
Add to cart
Product Type:
Purple and Pink and Dark Blue and Light Blue
Price (MYR)
1,280.00
PK
Add to cart
  • Modern patented design cools faster in freezers and stays cold longer on the bench top
  • PCR™ Cooler maintains a sample temperature of 0 °C (with 2 hours pre-cooling at -20 °C) for over an hour
  • When the temperature ceiling of 6 °C has been exceeded, users are signaled by a contrasting color change of the cooler, or even a single well
  • Cools industry standard format 0.2 mL and 0.5 mL PCR™ tubes, strips and single format, as well as most 96 well plates, in both standard and low profile well shape
  • Better alternative then ice baths as dry incubation reduces the risk of contamination
  • Freeze cooler in an upright position enabling cold storage of tubes
  • Well design prohibits the freezing of samples
  • Fully sealed and sonic welded cooler keeps coolant contained offering worry free usage
  • Save space with compact footprint and stacking ability
  • The lid is ambidextrous; it can be switched to accommodate left or right-hand orientation and is detachable
  • Robust cooler made from engineered polypropylene and the lid is made from polycarbonate
  • US Patent Pending, Community Design
  • No: 004101756-0001-0002
  • Two-Year Warranty
Specifications
Thermal Accuracy±1°C
Temperature Range (Metric)0°C to 5°C
For Use With (Equipment)Shakers, Mixers, Vortexers
CapacityTubes
Product TypePurple and Pink and Dark Blue and Light Blue
Height (Metric)1.9 cm
Width (Metric)14.8 cm
Depth (Metric)4.8 cm
For Use With (Application)PCR Applications
DescriptionFisherbrand™ PCR™ Cooler, 96-Well, 2 Pack
Quantity2
ColorPurple, Pink, Dark Blue, Light Blue
Vessel TypePCR Rack
Weight.27 kg (0.6 lb.)
Unit SizePK
Contents & Storage
Lid, Rack