PTFE Magnetic Stir Bars with Pivot Ring
PTFE Magnetic Stir Bars with Pivot Ring
Actual product may vary
PTFE Magnetic Stir Bars with Pivot Ring
Used in containers with bases that are slightly curved or uneven.
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)
FKS162558118 mm50 mm
Catalog number FKS16255811
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
8 mm
Length (Metric):
50 mm
  • Used in containers with bases that are slightly curved or uneven
Specifications
AutoclavableAutoclavable
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Diameter (Metric)8 mm
Length (Metric)50 mm
Quantity1/Pk.
ShapeCylinder
Unit SizePK