Clear Straight Sided Glass Jars with White Polypropylene Caps
Clear Straight Sided Glass Jars with White Polypropylene Caps
Actual product may vary
Clear Straight Sided Glass Jars with White Polypropylene Caps
Clear, USP Type III soda-lime glass wide mouth design is great for large solid samples and allow for complete removal of contents. Ideal for soil sampling and environmental applications
Have Questions?
Catalog NumberCapacity (Metric)Closure LinerQuantity
FKS1619450730 mLPTFE/Polyethylene Foam48
Catalog number FKS16194507
Price (MYR)
1,014.00
PK
Add to cart
Capacity (Metric):
30 mL
Closure Liner:
PTFE/Polyethylene Foam
Quantity:
48
Price (MYR)
1,014.00
PK
Add to cart
Clear, USP Type III soda-lime glass wide mouth design is great for large solid samples and allow for complete removal of contents. Ideal for soil sampling and environmental applications
Straight sided walls allow for the complete removal of contentsAvailable in a wide variety of sizes, from 1 to 32 oz.
Specifications
Capacity (Metric)30 mL
Capacity (English)1 oz.
ColorClear
Diameter (Metric) Outer Neck40 mm
Diameter (Metric) Inner Neck28 mm
For Use With (Application)Soil Sampling and Environmental Applications
IncludesCap
Height (Metric) With Closure46 mm
MaterialGlass
Quantity48
Closure ColorWhite
Neck Finish43-400
ClarityClear
Certifications/ComplianceUSP Type III Soda Lime Glass Wide Mouth Design is Great for Large Solid Samples and Allow for Complete Removal of Contents
Closure IncludedYes
MouthWide
ShapeRound
Height (Metric)43 mm
Product TypeStraight-sided Jar
Closure LinerPTFE/Polyethylene Foam
Closure MaterialPolypropylene
Closure Size43-400 mm
Diameter (Metric)43 mm
Closure TypeCap
Unit SizePK