Clear Boston Round Glass Bottles with Black Phenolic Caps
Clear Boston Round Glass Bottles with Black Phenolic Caps
Actual product may vary
Fisherbrand
Clear Boston Round Glass Bottles with Black Phenolic Caps
Clear Boston round Type III soda-lime glass is ideal for solvent, chemical or sample storage. Packaged in convenience packs with black phenolic caps attached to maintain cleanliness
Have Questions?
Catalog NumberCapacity (Metric)Closure Liner
FKS16154427240 mLRubber
Catalog number FKS16154427
Price (MYR)
183.00
PK
Add to cart
Capacity (Metric):
240 mL
Closure Liner:
Rubber
Price (MYR)
183.00
PK
Add to cart
Clear Boston round Type III soda-lime glass is ideal for solvent, chemical or sample storage. Packaged in convenience packs with black phenolic caps attached to maintain cleanliness
  • General use bottles ideal for when content integrity and sample or product visibility is important
  • Narrow screw neck opening, curved shoulders, round body
  • Glass offers minimal chemical interference with contents
  • Clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary
  • Environmentally sensitive - help eliminate waste and insure product integrity for long term storage
Specifications
Capacity (Metric)240 mL
Capacity (English)8 oz.
ColorClear
Diameter (Metric) Outer Neck21 mm
Diameter (Metric) Inner Neck13 mm
Height (Metric) With Closure139 mm
MaterialGlass (Type 3)
Quantity12
Closure ColorBlack
Neck Finish24-400
Closure IncludedYes
MouthNarrow
Height (Metric)136 mm
Product TypeBoston Round Bottle
Closure LinerRubber
Closure MaterialPhenolic
Closure Size24-400 mm
Diameter (Metric)60 mm
Unit SizePK