Clear Graduated Glass Bottles with White Polypropylene Caps
Clear Graduated Glass Bottles with White Polypropylene Caps
Actual product may vary
Fisherbrand
Clear Graduated Glass Bottles with White Polypropylene Caps
Fisherbrand™ Clear Graduated Glass Bottles with White Polypropylene Caps feature a wide mouth for easy removal of contents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
FKS16154377240 mL24
FISFB02911453A30 mL48
Catalog number FKS16154377
Price (MYR)
477.00
PK
Add to cart
Capacity (Metric):
240 mL
Quantity:
24
Price (MYR)
477.00
PK
Add to cart
  • Graduated in both milliliters and ounces
  • Clear glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications
Capacity (Metric)240 mL
Capacity (English)8 oz.
ColorClear
Diameter (Metric) Outer Neck54 mm
Diameter (Metric) Inner Neck40 mm
Height (Metric) With Closure130 mm
MaterialGlass (Type 3)
Quantity24
Closure ColorWhite
Neck Finish58-400
Closure IncludedYes
GraduatedYes
MouthWide
Height (Metric)127 mm
Product TypeGraduated Bottle
Closure LinerPulp/Vinyl
Closure MaterialPolypropylene
Closure Size58-400 mm
Diameter (Metric)61 mm
Unit SizePK