Nunc™ EasYDish™ Cell Culture Dishes, TC Surface
Nunc™ EasYDish™ Cell Culture Dishes, TC Surface
Actual product may vary
Nunc™ EasYDish™ Cell Culture Dishes, TC Surface
Innovative cell culture dishes uniquely designed to promote healthy cell growth and improve handling, stacking, and transporting of tissue cultures in the lab.
Have Questions?
Catalog NumberDiameter (Metric)Height (Metric)Culture Area
FKS15893937100 mm21 mm56.7 sq. cm
Catalog number FKS15893937
Price (MYR)
2,517.00
PK
Add to cart
Diameter (Metric):
100 mm
Height (Metric):
21 mm
Culture Area:
56.7 sq. cm
Price (MYR)
2,517.00
PK
Add to cart
Thermo Scientific Nunc EasYDish dishes are the next generation of cell culture dishes that improve handling, stacking, and transporting of cell cultures in your lab. The beveled grip ring is easy to hold and stays secure in your gloved hand. The dishes receive the standard Nunclon Delta surface treatment to maximize adhesion for the majority of cell types.
Beveled grip ring
  • Ensures lid stays in position when dish is being handled
  • Makes it easier to grasp and manage with a gloved hand
Proprietary notch design
  • Maximizes ventilation while maintaining sterility
  • Restricts the movement of the lid ensuring secure lid-to-dish fitting
  • Increases the physical strength of the dish
Raised outer edge on dish lid
  • Allows stabled stacking during transportation of cell cultures
Orientation marks on dish bottom
  • Simplifies locating of cells under the microscope
Writing area
  • Improves traceability of cell cultures
Re-sealable packaging
  • Promotes cleanliness of opened packs
Additional information
  • Material: polystyrene, USP VI
  • Contaminant free: RNase/DNase free, human DNA free, non-cytotoxic, non-pyrogenic
  • Sterility: gamma irradiation to SAL of 10-6
  • Certificate of Quality in every case
  • Surface treatment: Nunclon Delta for adherent cell cultures
Specifications
Certifications/ComplianceNunclon Delta certification
Closure IncludedClosure Included
Closure MaterialPolystyrene
DescriptionNunc EasYDish
AutoclavableNot Autoclavable
Bottom ShapeFlat
Capacity (Metric)12.5 mL (Well)
ColorClear
Culture Area56.7 sq. cm
DisposableReusable
GriddedNot Gridded
MaterialPolystyrene
ShapeRound
SterilitySterile
Surface TreatmentNunclon Delta
TypeCell Culture Dish
Volume (Metric) Working12.5 mL
Well Count1
For Use With (Application)Cell Culture
IncludesLid
No. per Case240/Cs.
No. per Pack10 Pk.
Shelf Life5 Years
Temperature Range (Metric)37°C
Height (Metric)21 mm
Diameter (Metric)100 mm
Unit SizePK