PowerPro 300
PowerPro 300
Actual product may vary
PowerPro 300
The PowerPro 300 is a versatile multi-function power supply suitable for a wide range of electrophoresis applications. At 300V, 700mA and 150W the PowerPro 300 has nearly twice the current and power of the market leading equivalent unit.
Have Questions?
Catalog NumberVoltage
FKS15818481100-240 V
Catalog number FKS15818481
Price (MYR)
7,937.00
EA
Add to cart
Voltage:
100-240 V
Price (MYR)
7,937.00
EA
Add to cart
  • Suited for use with horizontal electrophoresis systems and omniPAGE Mini vertical electrophoresis systems
  • Built in pre-set protocols and allows users to create and save protocols for their own systems
  • A 2.4” TFT-LCD Screen displays the current running parameters
  • Allows the user to browse pre-set and saved protocols
  • Can be operated at constant voltage, constant current or constant power and can store up to 30 programmes with up to 6 steps each
  • Convenient pause/resume key allows any run to be temporarily halted for inspection of the gel and restarted from the same position
Specifications
Current1 to 700 mA / 1 mA
For Use With (Application)Routine electrophoresis applications
TimerConstant mode: 9999 (min.) with alarm Programmable mode: 999 (min.) with alarm
TypePower Supply
Display2.4 in. TFT-LCD
Max. Voltage300 V
Voltage100-240 V
Unit SizeEA