Pour-Boat Weighing Dishes
Pour-Boat Weighing Dishes
Actual product may vary
Fisherbrand
Pour-Boat Weighing Dishes
Provide accurate pour-outs with minimal sample loss. Pour-Boat Weighing Dishes are polystyrene boats featuring smooth surfaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)
FKS1579817743 x 58 x 13 mm
FKS1570818783 x 132 x 26 mm
Catalog number FKS15798177
Price (MYR)
530.00
PK
Add to cart
Dimensions (L x W x H):
43 x 58 x 13 mm
Price (MYR)
530.00
PK
Add to cart
  • Pour-spout design easily dispenses samples without flexing the dish.
  • Flat-bottom dishes are made from 12-mil thick polystyrene for better strength, stability, and handling.
  • Ideal for solid product weighing.
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • Temperature range -10°C to +70°C (14°F to 158°F).
  • 1 color 3 sizes.
  • Can be used with microwave moisture analyzers.
  • FDA/food-grade compliant.
  • Pack of 500
Specifications
AutoclavableNot Autoclavable
Bottom ShapeFlat
ColorWhite
MaterialPolystyrene
ShapeBoat
SterilityNon-sterile
TypeWeighing Dish
For Use With (Equipment)Microwave Moisture Analyzer
Quantity500 Pk.
Temperature Range (English)14°F to 158°F
Temperature Range (Metric)-10°C to +70°C
Thickness12 mil
Dimensions (L x W x H)43 x 58 x 13 mm
Length (English)1.7 in.
Length (Metric)43 mm
Width (English)2.3 in.
Width (Metric)58 mm
Height (Metric)13 mm
Height (English)0.5 in.
Unit SizePK