Digital Vortex Mixer
Digital Vortex Mixer
Actual product may vary
Fisherbrand
Digital Vortex Mixer
Ideal for applications that demand repeatable results.
Have Questions?
Catalog NumberVoltageAmperage
FKS15796959230 V0.13 A
Catalog number FKS15796959
Price (MYR)
3,935.00
EA
Add to cart
Voltage:
230 V
Amperage:
0.13 A
Price (MYR)
3,935.00
EA
Add to cart
Microprocessor control maintains set point for strong, consistent mixing action. Timer will display elapsed time or, when programmed to user defined time limit, the unit will shut off when the time reaches zero. Two modes of operation: “Continuous” mode when using accessory attachments or “Touch” mode which activates mixing when depressing the cup head.
  • LED displays for speed and time
  • Continuous or touch modes
  • Microprocessor controls
  • 5 year warranty
Specifications
Orbit4.9 mm
Speed Range500 to 3000 rpm
Electrical Requirements230 V, 0.13 A, 135 W
Voltage230 V
Wattage135 W
Amperage0.13 A
Product TypeDigital Vortex Mixer
Height (English)5 in.
Height (Metric)12.7 cm
Length (English)8in.
Length (Metric)20.2 cm
Width (English)4.75 in.
Width (Metric)12.1 cm
Shipping Weight (English)10 lb.
Shipping Weight (Metric)5.3 kg
Weight (English)10 lb.
Weight (Metric)4.5 kg
IncludesIncludes both cup head and 3 in. head with cover. Units are supplied with a 72 in. (183cm), 3-wire cord and plug.
Warranty and Service Offering5 year limited warranty on parts and labor.
Unit SizeEA