Square Weighing Boats
Square Weighing Boats
Actual product may vary
Square Weighing Boats
Square Weighing Boats
Square Weighing Boats
Provide accurate pour-outs with minimal sample loss. Square Weighing Boats are flexible polystyrene boats featuring smooth surfaces.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1575818751 mL
Catalog number FKS15758187
Price (MYR)
578.00
PK
Add to cart
Capacity (Metric):
51 mL
Price (MYR)
578.00
PK
Add to cart
  • Shallow boat with a wide, flat bottom to resist tipping.
  • Ideal for solid product weighing.
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • Temperature range -10 to 70°C (14 to 158°F).
  • Can be used with microwave moisture analyzers.
  • FDA/food-grade compliant.
  • Pack of 500
  • Disposable
Specifications
Capacity (English)1.7 oz.
AutoclavableNot Autoclavable
Bottom ShapeFlat
Capacity (Metric)51 mL
ColorWhite
DisposableSingle-use
MaterialPolycarbonate
ShapeSquare
SterilityNon-sterile
TypeWeighing Boat
For Use With (Equipment)Microwave Moisture Analyzer
Dimensions (L x W x H)90 x 90 x 7 mm
Length (English)3.5 in.
Length (Metric)90 mm
Width (English)3.5 in.
Width (Metric)90 mm
Height (Metric)7 mm
Height (English)0.3 in.
Unit SizePK