4-Watt UV Lamp
4-Watt UV Lamp
Actual product may vary
4-Watt UV Lamp
These handheld lamps come in a variety of wavelengths and are ideal for viewing materials in dimly-lit areas. The different wavelengths can be used for bacterial identification, specimen staining, biochemical testing, mercury detectors, optical alignment, DNA analysis, electrophoresis and much more.
Have Questions?
Catalog number FKS15651874
Price (MYR)
-
  • Easily switch from one UV light source to another
  • Built-in reflector enhances UV illumination
  • Durable, extruded metal housing features scratch-resistant powder coating
  • Lightweight and easy to handle
  • Offers two to three different UV wavelengths in one lamp
  • Supplied with 254 nm and 365 nm wavelenth tubes
  • Selector switch makes it easy to switch between light sources
  • 365 nm longwave UV models are ideal for bacterial identification, specimen staining, gel electrophoresis, chlorination, fluorochemistry and more
  • 365 nm longwave UV models are ideal for bacterial identification, specimen staining, gel electrophoresis, chlorination, fluorochemistry and more
  • 254 nm shortwave UV models can be used for biochemical testing, mercury detectors, optical alignment, DNA analysis, electrophoresis and more
  • Optional viewing cabinets available
  • Optional lamp stand for hands-free operation is availalbe for all models
Specifications
Voltage230 V
Wattage4 W
Dimensions (L x W)24.8 x 6.4 cm
Includes254 nm and 365 nm wavelength tubes
Lighting Intensity590/900 μ/cm2
Unit SizeEA