Microcentrifuge Tube Racks
Microcentrifuge Tube Racks
Actual product may vary
Microcentrifuge Tube Racks
Accommodates 0.5ml microcentrifuge tubes on one side while the opposite side holds 1.5ml to 2.0ml microcentrifuge tubes. 96 wells on each side keep tubes upright in an 8 x 12 array.
Have Questions?
Catalog NumberColor
FKS15550062Natural
Catalog number FKS15550062
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Natural
  • Rack colors assist in sample organization and quick visual identification
  • Alphanumeric index permits easy tube identification
  • Matte side panel accepts tape or direct labeling
  • Molded in handles provide a secure grip for transport
  • Can be used for freezer storage down to -80°C (-112°F)
  • Autoclavable at 121°C (250°F)
  • Overall dimensions (L x W x H): 21.3 x 11.4 x 2.7cm (8-3/8 x 4-1/2 x 1-1/16 in.)
Specifications
IncludesMatte Side Panel for Labeling
Product TypeMicrotube Rack
Closure IncludedNo
Array8 x 12
AutoclavableAutoclavable (121°C)
ColorNatural
HoldsMicrotubes (96)
MaterialPolypropylene
Positions96
ShapeRectangular
For Use With (Equipment)1.5/2.0mL microtubes
Quantity5/Pk.
Temperature (Metric) Range-80°C
Dimensions (L x W x H)213 x 114 x 27mm
Length (Metric)213 mm
Width (Metric)114 mm
Height (Metric)27 mm
Unit SizePK