Squat Form Beakers
Squat Form Beakers
Actual product may vary
Fisherbrand
Squat Form Beakers
Constructed of borosilicate glass. Fisherbrand™ Squat Form Beakers are graduated and come in a large variety of capacities.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS15499073150 mL
FKS1546907325 mL
FKS1547907350 mL
FKS15489073100 mL
FKS15409083250 mL
FKS15419083400 mL
FKS15429083600 mL
FKS154490831000 mL
FKS154590832000 mL
FKS154790835000 mL
Catalog number FKS15499073
Price (MYR)
199.00
PK
Add to cart
Capacity (Metric):
150 mL
Price (MYR)
199.00
PK
Add to cart
Constructed of borosilicate glass. Fisherbrand™ Squat Form Beakers are graduated and come in a large variety of capacities.
Fisherbrand™ borosilicate glass beakers. Squat form with spout and graduations. ISO 3819 DIN 12331.
Specifications
ColorTranslucent
Diameter (Metric) Outer60 mm
Capacity (Metric)150 mL
MaterialBorosilicate Glass
FinishMatte
Height (Metric)80 mm
IncludesSpout
Quantity10 Pk.
TypeBeaker
GraduatedYes
Certifications/ComplianceISO 3819, DIN 12331
ScaleWhite Graduated Scale
StyleLow Form
Unit SizePK