LO-tox™ Lab Thermometer
LO-tox™ Lab Thermometer
Actual product may vary
LO-tox™ Lab Thermometer
LO-tox™ Lab Thermometer
Fisherbrand
LO-tox™ Lab Thermometer
LO-tox™ lab thermometer without calibration certificate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature Range (Metric)SubdivisionImmersion TypeLength (Metric)
FKS15432695-10°C to +360°C2°CTotal Immersion305 mm
FKS13058603-10°C to +101°C0.2°CPartial Immersion405 mm
Catalog number FKS15432695
Price (MYR)
711.00
PK
Add to cart
Temperature Range (Metric):
-10°C to +360°C
Subdivision:
2°C
Immersion Type:
Total Immersion
Length (Metric):
305 mm
Price (MYR)
711.00
PK
Add to cart
Read temperatures quickly and accurately with blue spirit against a white background. Brannan™ Lo-Tox™ Spirit-filled partial Immersion Thermometer is manufactured to traceable standards and includes a blue non-roll top and safety suspension ring.
  • General purpose/stirring thermometer with a low toxic, environmentally friendly filling
  • Easy to read against background
  • Each thermometer is manufactured to traceable standards
Specifications
Immersion TypeTotal Immersion
Length (Metric)305 mm
MaterialGlass
Thermometer FillBrown Organic Filling
Product TypeLaboratory Immersion Thermometer
Max. Temperature (Metric)360°C
Min. Temperature (Metric)-10°C
Temperature Range (Metric)-10°C to +360°C
Accuracy±2°C at 90°C
Subdivision2°C
Unit SizePK