Isotemp™ Hotplate Stirrer, Ambient to 400°C, Ceramic
Isotemp™ Hotplate Stirrer, Ambient to 400°C, Ceramic
Actual product may vary
Isotemp™ Hotplate Stirrer, Ambient to 400°C, Ceramic
Isotemp™ Hotplate Stirrer, Ambient to 400°C, Ceramic
Isotemp™ Hotplate Stirrer, Ambient to 400°C, Ceramic
Isotemp™ Hotplate Stirrer, Ambient to 400°C, Ceramic
Stirring Range: 50 to 1500 rpm, Shape: Square, Size: 10.25 x 10.25 in., Plug Type: EU, UK, CN
Have Questions?
Catalog NumberPlug Type
FKS15353538UK/EU/CN
Catalog number FKS15353538
Price (MYR)
6,473.00
EA
Add to cart
Plug Type:
UK/EU/CN
Price (MYR)
6,473.00
EA
Add to cart
Experience precise stirring control, exceptional safety and temperature performance for your routine protocols.
Advanced technology:
  • Slow-speed stirring, consistent speed control, and strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design:
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
  • Includes stir bar
  • Plug: EU, UK, CN
Specifications
Top Plate MaterialCeramic
Voltage230 V
DescriptionHotplate Stirrer
IncludesStir Bar
Electrical Requirements230 V 50/60 Hz
Length (English)11.3 in.
Size10.25 x 10.25 in.
Height (Metric)10.2 cm
Temperature (Metric)Ambient to 400°C
Height (English)4 in.
Frequency50/60 Hz
Width (Metric)41.1 cm
Length (Metric)28.7 cm
Width (English)16.2 in.
TypeStirring Hotplate
ShapeSquare
Plug TypeUK/EU/CN
Max. Container Size6 L
Stirring Range50 to 1500 rpm
Warranty3 Years
Dimensions (LxWxH)10.25 in. x 10.25 in.
Certifications/ComplianceRoHS, cCSAus (100 to 120 V Model), CE (230 V Model)
Unit SizeEA