Check Order Status
Low Form Polypropylene Graduated Beakers
Actual product may vary
Fisherbrand
Low Form Polypropylene Graduated Beakers
With good temperature and chemical resistance, these beakers are excellent for general laboratory use.
Have Questions?
Catalog Number
Capacity (Metric)
Scale
FKS15330454
250 mL
Blue Printed Scale
Catalog number
FKS15330454
Price (MYR)
195.00
PK
Add to cart
Capacity (Metric):
250 mL
Scale:
Blue Printed Scale
Price (MYR)
195.00
PK
Add to cart
With graduation marks in either blue or embossed, beaded rim and spout
Cleaning at temperatures below 60°C is recommended to preserve marks and inscriptions
For autoclaving at 121°C, beakers with an embossed scale should be selected
Specifications
Color
High clarity
Diameter (Metric) Outer
77 mm
Capacity (Metric)
250 mL
Material
Polypropylene
Height (Metric) Outer
95 mm
Increments
25 mL
Quantity
6/Pk.
Type
Beaker
Autoclavable
Autoclavable
Graduated
Yes
Scale
Blue Printed Scale
Style
Low Form