Flatpack Freezer Boxes
Flatpack Freezer Boxes
Actual product may vary
Flatpack Freezer Boxes
Outlasts carboard alternatives and reduces the chance of mould growth. Fisherbrand™ Flatpack Freezer Boxes can freeze and thaw multiple times without a problem.
Have Questions?
Catalog NumberColorHoldsTube ArrayDimensions (L x W x H)
FKS15316528Natural144 (0.2 mL Tubes)12 x 12133 x 130 x 30 mm
Catalog number FKS15316528
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Natural
Holds:
144 (0.2 mL Tubes)
Tube Array:
12 x 12
Dimensions (L x W x H):
133 x 130 x 30 mm
  • Cryoboxes are supplied flat-pack eliminating the hassle of stacking and reduces the amount of space needed for storage
  • Boxes fit in standard freezer racks
  • Easy to assemble
  • Constructed from corrugated polypropylene
  • More durable than cardboard boxes
  • Not susceptible to moisture
  • Significantly reduces the chance of mould
  • Dividers included with the boxes
  • Store down to temperatures of -80°C
Specifications
Closure TypeLid
ColorNatural
MaterialPolypropylene
ShapeRectangular
TypeFreezer Box
Holds144 (0.2 mL Tubes)
IncludesLid and Dividers
Quantity10 Pack
Tube Array12 x 12
Dimensions (L x W x H)133 x 130 x 30 mm
Length (Metric)133 mm
Width (Metric)130 mm
Height (Metric)30 mm
Diameter (Metric) Well7.9 mm
Weight (Metric)0.57 kg
Unit SizePK